Learn More
Medchemexpress LLC Nonanoic acid-d4 | 1219795-27-3 | 1 MG
SDP
Catalog No. 5000419627
Encompass_Preferred
Nonanoic acid-d4, also known as Pelargonic acid-d4, is a deuterium-labeled version of Nonanoic acid. Nonanoic acid is a naturally occurring saturated fatty acid composed of nine carbon atoms. This compound is known to significantly decrease bacterial translocation, boost antibacterial activity, and notably increase the production of porcine β-defensins 1 (pBD-1) and pBD-2.
  • Stable isotope-labeled compound.
  • Can be utilized as a tracer in various research applications.
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Valuable for pharmacokinetic and metabolic studies due to the potential of deuteration to influence drug profiles.
  • Significantly reduces bacterial translocation.
  • Enhances antibacterial activity.
  • Remarkably increases the secretion of porcine β-defensins 1 (pBD-1) and pBD-2.
Catalog No. 50-004-19627 Supplier Medchemexpress LLC Supplier No. HYN7057S21MG
Nonanoic acid-d4, also known as Pelargonic acid-d4, is a deuterium-labeled version of Nonanoic acid. Nonanoic acid is a naturally occurring saturated fatty acid composed of nine carbon atoms. This compound is known to significantly decrease bacterial translocation, boost antibacterial activity, and notably increase the production of porcine β-defensins 1 (pBD-1) and pBD-2.
  • Stable isotope-labeled compound.
  • Can be utilized as a tracer in various research applications.
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Valuable for pharmacokinetic and metabolic studies due to the potential of deuteration to influence drug profiles.
  • Significantly reduces bacterial translocation.
  • Enhances antibacterial activity.
  • Remarkably increases the secretion of porcine β-defensins 1 (pBD-1) and pBD-2.