Learn More
Medchemexpress LLC Nonanoic acid-d4 | 1219795-27-3 | 5 MG
SDP
Catalog No. 5000418680
Encompass_Preferred
Nonanoic acid-d4 is the deuterium labeled Nonanoic acid, a naturally-occurring saturated fatty acid with nine carbon atoms. It significantly reduces bacterial translocation, enhances antibacterial activity, and remarkably increases the secretion of porcine β-defensins 1 (pBD-1) and pBD-2.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-18680 Supplier Medchemexpress LLC Supplier No. HYN7057S25MG
Nonanoic acid-d4 is the deuterium labeled Nonanoic acid, a naturally-occurring saturated fatty acid with nine carbon atoms. It significantly reduces bacterial translocation, enhances antibacterial activity, and remarkably increases the secretion of porcine β-defensins 1 (pBD-1) and pBD-2.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.