Learn More
Medchemexpress LLC Nonanoic acid-d4 (Pelargonic acid-d4) | 1219795-27-3 | 10 MG
SDP
Catalog No. 5000419673
Encompass_Preferred
Nonanoic acid-d4 is the deuterium labeled Nonanoic acid, a naturally-occurring saturated fatty acid with nine carbon atoms. It significantly reduces bacterial translocation, enhances antibacterial activity, and remarkably increases the secretion of porcine β-defensins 1 (pBD-1) and pBD-2. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Reduces bacterial translocation
  • Enhances antibacterial activity
  • Increases secretion of porcine β-defensins 1 (pBD-1) and pBD-2
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development
Catalog No. 50-004-19673 Supplier Medchemexpress LLC Supplier No. HYN7057S210MG
Nonanoic acid-d4 is the deuterium labeled Nonanoic acid, a naturally-occurring saturated fatty acid with nine carbon atoms. It significantly reduces bacterial translocation, enhances antibacterial activity, and remarkably increases the secretion of porcine β-defensins 1 (pBD-1) and pBD-2. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Reduces bacterial translocation
  • Enhances antibacterial activity
  • Increases secretion of porcine β-defensins 1 (pBD-1) and pBD-2
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development