Learn More
Medchemexpress LLC Nonanoic acid, methyl ester | 1731-84-6 | MFCD00009569 | 98.7% | 172.27 | C10H20O2 | 10 G
SDP
Catalog No. 5000302410
Encompass_Preferred
Methyl nonanoate (nonanoic acid methyl ester) is a colourless liquid methyl ester with a fruity aroma used in flavor and fragrance applications, as an analytical standard, and as an intermediate in chemical manufacturing. It is employed in formulations, perfumery, and industrial synthesis where a medium-chain ester is required.
  • Fruity odor suitable for flavor and fragrance formulations.
  • High purity appropriate for analytical and research use.
  • Useful intermediate for synthesis of solvents, resins, and plasticizers.
  • Liquid at room temperature for easy handling and dispensing.
  • Accompanied by safety and quality documentation (SDS, COA).
Catalog No. 50-003-02410 Supplier Medchemexpress LLC Supplier No. HYW01530810G
Methyl nonanoate (nonanoic acid methyl ester) is a colourless liquid methyl ester with a fruity aroma used in flavor and fragrance applications, as an analytical standard, and as an intermediate in chemical manufacturing. It is employed in formulations, perfumery, and industrial synthesis where a medium-chain ester is required.
  • Fruity odor suitable for flavor and fragrance formulations.
  • High purity appropriate for analytical and research use.
  • Useful intermediate for synthesis of solvents, resins, and plasticizers.
  • Liquid at room temperature for easy handling and dispensing.
  • Accompanied by safety and quality documentation (SDS, COA).