Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonanoic Acid 90.0+%, TCI America™
SDP

Catalog No. N0288500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0288500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0288500ML Supplier TCI America Supplier No. N0288500ML

missing translation for 'validMainframeMsg'

Specifications

CAS 112-05-0
Color Yellow
Boiling Point 254°C
Molecular Formula C9H18O2
MDL Number MFCD00004433
UN Number 3265
Quantity 500 mL
Synonym pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a
InChI Key FBUKVWPVBMHYJY-UHFFFAOYSA-N
SMILES CCCCCCCCC(=O)O
IUPAC Name nonanoic acid
Molecular Weight (g/mol) 158.241
PubChem CID 8158
ChEBI CHEBI:29019
Formula Weight 158.24
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonanoic Acid
Show More Show Less
EINECSNumber (2)-0608
RTECSNumber RA6650000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.