Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonanophenone 97.0+%, TCI America™
SDP

Catalog No. N056825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N056825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N056825ML Supplier TCI America Supplier No. N056825ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6008-36-2
Molecular Formula C15H22O
Molecular Weight (g/mol) 218.34
MDL Number MFCD00048965
InChI Key PFUPABFCHVRLLY-UHFFFAOYSA-N
Synonym nonanophenone, n-nonanophenone, n-octyl phenyl ketone, 1-phenyl-1-nonanone, 1-nonanone, 1-phenyl, 1-phenyl-nonan-1-one, acmc-209mhy
PubChem CID 80108
IUPAC Name 1-phenylnonan-1-one
SMILES CCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 177°C
Quantity 25 mL
Formula Weight 218.34
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonanophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.