Learn More
Medchemexpress LLC Nonanoyl-CoA-d17 | >98.0% | 924.86 | C30H35D17N7O17P3S | 1mg
SDP
Supplier:  Medchemexpress LLC HYW7062341MG
Encompass_Preferred
Nonanoyl-CoA-d17 (Pelargonoyl-CoA-d17) is the deuterium labeled Nonanoyl-CoA (HY-E70246) Nonanoyl-CoA is a medium-chain fatty acyl-CoA that can be used as substrate for the medium-chain hydrolase[1]
Catalog No. 50-002-70164