Learn More
Medchemexpress LLC HY-P0097A 1mg Medchemexpress, Nonapeptide-1 acetate salt CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0097A1MG
Encompass_Preferred
Medchemexpress, HY-P0097A 1mg Nonapeptide-1 acetate salt CAS: Nonapeptide-1 acetate salt, a peptide hormone, is a potent α-Melanocyte-stimulating hormone (α-MSH) antagonist, with an IC 50 of 11 nM. Reduces synthesis of melanin and helps decrease skin pigmentation to a substantial degree [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4020