Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonasaccharide Glc4Xyl3Gal2 75.0+%, TCI America™
SDP

Catalog No. N0693100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0693100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0693100MG Supplier TCI America Supplier No. N0693100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 129865-06-1
Molecular Formula C51H86O43
Molecular Weight (g/mol) 1387.206
MDL Number MFCD04040971
InChI Key GSCHIGXDTVYEEM-UHFFFAOYSA-N
Synonym XLLG, Xyloglucan Nonasaccharide
PubChem CID 44630412
IUPAC Name 2-[2-[[3-[6-[[4,5-dihydroxy-3-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]oxymethyl]-3,4-dihydroxy-5-[3,4,5-trihydroxy-6-[(3,4,5-trihydroxyoxan-2-yl)oxymethyl]oxan-2-yl]oxyoxan-2-yl]oxy-4,5-dihydroxy-6-[4,5,6-trihydroxy-2-(hydroxymethyl)oxan
SMILES C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)O)CO)COC6C(C(C(CO6)O)O)OC7C(C(C(C(O7)CO)O)O)O)COC8C(C(C(CO8)O)O)OC9C(C(C(C(O9)CO)O)O)O)O)O)O)O)O)O

Specifications

Color White
Quantity 100 mg
Formula Weight 1387.21
Percent Purity ≥75.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nonasaccharide Glc4Xyl3Gal2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.