Learn More
Cayman Chemical Neoandrographolide, Purity: ≥95%, CAS: 27215-14-1, 10 mg
SDP
Catalog No. 502951955
Encompass_Preferred
A principle diterpenoid isolated from A. paniculata that inhibits LPS-induced nitric oxide production in inflammatory macrophages after oral administration to mice (25 mg/kg) or by direct addition to cultured macrophages (IC50 = 35.5 µM); reduces VEGF-induced proliferation of HUVECs; inhibits furin with an IC50 value of 53.5 µM
Catalog No. 50-295-1955 Supplier Cayman Chemical Supplier No. 1174210 MG
A principle diterpenoid isolated from A. paniculata that inhibits LPS-induced nitric oxide production in inflammatory macrophages after oral administration to mice (25 mg/kg) or by direct addition to cultured macrophages (IC50 = 35.5 µM); reduces VEGF-induced proliferation of HUVECs; inhibits furin with an IC50 value of 53.5 µM