Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonenylsuccinic anhydride, 96%

Catalog No. AAA1760718
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1760718 50 g
AAA1760730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1760718 Supplier Thermo Scientific Chemicals Supplier No. A1760718
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 28928-97-4 | C13H20O3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28928-97-4
Molecular Formula C13H20O3
MDL Number MFCD00014552

Specifications

Density 1.032
Boiling Point 126°C to 130°C (0.15 mmHg)
Flash Point 146°C (295°F)
Quantity 50 g
Sensitivity Moisture sensitive
Formula Weight 224.3
Percent Purity 96%
Chemical Name or Material Nonenylsuccinic anhydride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 249-317-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.