Learn More
Crescent Chemical Co Inc NONENYLSUCCINIC ANHYDRIDE PURE
SDP
Supplier:  Crescent Chemical Co Inc SE30812.01
Encompass_Preferred
Nonenylsuccinic anhydride pure Especially purified for electron microscopy. Hardener component for SPURR embedding. Size - 250ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 30812.01 CAS 28928-97-4
Catalog No. 50-247-947