Learn More
ATCC Non-Enzymatic Cell Dissociation Media, 1X, 100 ML
Catalog No. 502383426
Encompass
Trypsin is the most commonly used enzyme for harvesting cells in culture. A non-enzymatic approach is needed when non-protein and animal-component free materials are required. ATCC Non-Enzymatic Cell Dissociation Solution is a sterile, phenol-red free solution composed of a proprietary mixture of chelators representing an optimized alternative to protein-digesting enzymes. This product is totally free of animal-derived components. This product has applications for cell culture, cell growth, and viability.
Catalog No. 50-238-3426 Supplier ATCC Supplier No. 302103
Trypsin is the most commonly used enzyme for harvesting cells in culture. A non-enzymatic approach is needed when non-protein and animal-component free materials are required. ATCC Non-Enzymatic Cell Dissociation Solution is a sterile, phenol-red free solution composed of a proprietary mixture of chelators representing an optimized alternative to protein-digesting enzymes. This product is totally free of animal-derived components. This product has applications for cell culture, cell growth, and viability.