Learn More
Quality Biological Inc Instant Non-Fat Dry Milk 3 Quart
SDP
Catalog No. NC2337290
Encompass_Preferred
Nonfat Dry Milk is often used in blocking solutions to reduce background and minimize non-specific binding. Nonfat Dry Milk is often a preferred component of Blocking Solutions because it is inexpensive and widely available.
Can be used for making blocking solution
Blocking solutions decrease the non-specific binding of peptides and proteins to improve assay sensitivity.
Storage Conditions: 15 °C to 30 °C
Product and/or packaging contains recycled/renewable content Product and/or packaging is recyclable or reduces waste Product and/or packaging is safer to human and environmental health
Catalog No. NC2337290 Supplier Quality Biological Inc Supplier No. A6141001
Nonfat Dry Milk is often used in blocking solutions to reduce background and minimize non-specific binding. Nonfat Dry Milk is often a preferred component of Blocking Solutions because it is inexpensive and widely available.
Can be used for making blocking solution
Blocking solutions decrease the non-specific binding of peptides and proteins to improve assay sensitivity.
Storage Conditions: 15 °C to 30 °C
Product and/or packaging contains recycled/renewable content Product and/or packaging is recyclable or reduces waste Product and/or packaging is safer to human and environmental health