Learn More
Perkin Elmer US LLC Non-Flared Santoprene MP2 Two-Stop Peristaltic Pump Tubing - 1.52 mm I.D., Yellow/Blue, Pkg. 12
Catalog No. 502104993
Encompass_Preferred
This MP2 Two-Stop Peristaltic Pump Tubing is designed for use with the MP2 pump on your ICP-OES and ICP-MS instrument. The MP2 is a compact, precise, stand-alone micro peristaltic pump with a flow rate range from 0.20 µL/min to 10 mL/min and is available with one to eight channels. Optimized for low-flow rates, the MP2 uses either MP2 tubing or standard 3-stop pump tubing.
Catalog No. 50-210-4993 Supplier Perkin Elmer US LLC Supplier No. N8145174
This MP2 Two-Stop Peristaltic Pump Tubing is designed for use with the MP2 pump on your ICP-OES and ICP-MS instrument. The MP2 is a compact, precise, stand-alone micro peristaltic pump with a flow rate range from 0.20 µL/min to 10 mL/min and is available with one to eight channels. Optimized for low-flow rates, the MP2 uses either MP2 tubing or standard 3-stop pump tubing.