Learn More
Research Products International Corp Nonidet P-40 Substitute, 500 Milliliters
SDP
Catalog No. 50488856
Encompass_Preferred
Laboratory reagent grade. Nonidet P-40 is a nonionic, non-denaturing detergent.
Catalog No. 50-488-856 Supplier Research Products International Corp Supplier No. N59000500.0
Laboratory reagent grade. Nonidet P-40 is a nonionic, non-denaturing detergent.