Learn More
Hach Company Nonionic Surfactants TNTplus Vial Test, HR (6.0-200 mg/L as Triton X-100), 25 Tests
Catalog No. 501998160 Shop All Hach Company Products
Encompass
Nonionic Surfactants TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results everytime.
Catalog No. 50-199-8160 Supplier Hach Company Supplier No. TNT876
Nonionic Surfactants TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results everytime.