Learn More
Thermco Products Inc NONMERCURY THERMOMETER THERMCO
Catalog No. NC1146574
Encompass_Preferred
-100/50 x 1.0C, Total immersion, Spirit, Certified -80, -70, -20, 0, 37C, w/case
Catalog No. NC1146574 Supplier Thermco Products Inc Supplier No. ACC603XSCFC
-100/50 x 1.0C, Total immersion, Spirit, Certified -80, -70, -20, 0, 37C, w/case