Learn More
Aviva Systems Biology Corp NONO Antibody - C-terminal region (ARP40716_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40716_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NONO. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human NONO. Alternate names: P54, NMT55, NRB54, P54NRBApplicationIF, IHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53226