Learn More
Cayman Chemical Non-polar Lipid Mixture 2 (plant, ovine), 1 mL
SDP
Catalog No. 502990007
Encompass_Preferred
A mixture of non-polar lipids; intended for use as a reference standard for the detection of these non-polar lipids in various sample types by TLC or LC-MS
Catalog No. 50-299-0007 Supplier Cayman Chemical Supplier No. 293771 ML
A mixture of non-polar lipids; intended for use as a reference standard for the detection of these non-polar lipids in various sample types by TLC or LC-MS