Learn More
Covidien Nonsterile Monoject™ Syringes without Needles
Catalog No. 22257144
Change view
Click to view available options
Volume (Metric):
3 mL
6 mL
12 mL
Connection Type:
Regular
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Connection Type
22-257-144 6 mL Regular
22-257-143 3 mL Regular
22-257-145 12 mL Regular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-257-144 Supplier Covidien Supplier No. 8881106028
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Bulk packaged without needles, available in several sizes
  • Permanent heat-etched graduations allow precise fluid measurements
  • Self-sealing, latex-free plunger tip
Specifications
Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) Used as general purpose syringe
Latex Free Yes
Product Type Monoject™ Syringe
Needle Point Number No Needle Included
Volume (Metric) 6 mL
Connection Type Regular
Graduated Yes
Sterility Non-sterile