Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nonsterile Pipette Tips for Beckman™ Liquid Handling Systems

Catalog No. p-4525556
Change view
Click to view available options
Volume (Metric):
250 μL
For Use With (Equipment):
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX SPAN-8
Fit for Purpose:
Certified Quality
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) For Use With (Equipment) Fit for Purpose
21-402-524 250 μL Beckman Coulter Biomek FX, Beckman Coulter Biomek NX SPAN-8 Certified Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-402-524 Supplier Thermo Scientific™ Supplier No. 919252
This item is not returnable. View return policy

Increase pipetting performance through the superior tip straightness and low %CV of these pipette tips for automated liquid handling systems.

Increase pipetting performance with Thermo ScientificTM Nonsterile Pipette Tips for BeckmanTM Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. They provide accurate and precise performance because of superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.

Features:

  • Available in low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384- and 1536- DeepWell™ plates
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 250 μL
Filtered Non-filtered
Sterility Non-sterile
Format Rack
Tip Style Conductive
For Use With (Equipment) Beckman Coulter Biomek FX, Beckman Coulter Biomek NX SPAN-8
Product Line Beckman
Fit for Purpose Certified Quality
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.