Learn More
Electron Microscopy Sciences Pad-Pet Pipettes 1.0 ml, 25 µl drop, 13.1 cm
SDP
Catalog No. 501899405
Encompass_Preferred
These polyethylene disposable have a low-affinity surface, attached bulb avoids cross-contamination, and are one step biohazard disposable. These pipets are ideal for chemical sampling or transferring, smear blood sample onto slides, mix and spread reagent, slide tests, etc. Non-sterile only.
Size: 25 µl drop
OAL: 13.1 cm
Catalog No. 50-189-9405 Supplier Electron Microscopy Sciences Supplier No. 7096318
These polyethylene disposable have a low-affinity surface, attached bulb avoids cross-contamination, and are one step biohazard disposable. These pipets are ideal for chemical sampling or transferring, smear blood sample onto slides, mix and spread reagent, slide tests, etc. Non-sterile only.
Size: 25 µl drop
OAL: 13.1 cm