Learn More
Biotium Nonyl Acridine Orange (NAO)
Catalog No. 501964747
Encompass_Preferred
NAO is a mitochondrial dye whose staining is not dependent on mitochondrial membrane potential, unlike JC-1 and Rhodamine 123. NAO has been used to study multidrug resistance and to measure changes in mitochondrial mass during apoptosis.Properties: Ex/Em (MeOH) = 495/522nm; e (MeOH) = 63,000; Orange solid soluble in DMSO or DMF; Store at 4°C and protect from light, especially in solution; C26H38BrN3; MW: 473; [75168-11-5]
Catalog No. 50-196-4747 Supplier Biotium Supplier No. 70012
NAO is a mitochondrial dye whose staining is not dependent on mitochondrial membrane potential, unlike JC-1 and Rhodamine 123. NAO has been used to study multidrug resistance and to measure changes in mitochondrial mass during apoptosis.Properties: Ex/Em (MeOH) = 495/522nm; e (MeOH) = 63,000; Orange solid soluble in DMSO or DMF; Store at 4°C and protect from light, especially in solution; C26H38BrN3; MW: 473; [75168-11-5]