Learn More
Sigma Aldrich Fine Chemicals Biosciences Nonyl beta D glucopyranosi5G
Catalog No. 502914454
Encompass
Nonyl -D-glucopyranoside is a nonionic surfactant among a small number of surfactants found to be useful for the solubilization and crystallization of a large number of biological membrane proteins.
Catalog No. 50-291-4454 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N75075G
Nonyl -D-glucopyranoside is a nonionic surfactant among a small number of surfactants found to be useful for the solubilization and crystallization of a large number of biological membrane proteins.