Learn More
Medchemexpress LLC Nonyl β-D-glucopyranoside | 69984-73-2 | MFCD00063300 | 99.8% | 306.40 | C15H30O6 | 1 G
SDP
Catalog No. 5000302116
Encompass_Preferred
Nonyl β-D-glucopyranoside is a non-ionic alkyl-glucoside surfactant used in biochemical and surface-chemistry applications. It is commonly used to study interactions with lipid monolayers, to solubilize hydrophobic biomolecules, and to support chiral separations of certain acids and amino acid derivatives.
  • Non-ionic surfactant suitable for membrane and lipid monolayer studies.
  • Aids solubilization of membrane proteins and hydrophobic compounds.
  • Supports chiral separations of certain herbicides and amino acid derivatives.
  • High listed purity (≈99.8%) for reproducible results.
  • Molecular weight 306.40 and formula C15H30O6.
  • Available in solid and solution formats for flexible use.
Catalog No. 50-003-02116 Supplier Medchemexpress LLC Supplier No. HYW0159931G
Nonyl β-D-glucopyranoside is a non-ionic alkyl-glucoside surfactant used in biochemical and surface-chemistry applications. It is commonly used to study interactions with lipid monolayers, to solubilize hydrophobic biomolecules, and to support chiral separations of certain acids and amino acid derivatives.
  • Non-ionic surfactant suitable for membrane and lipid monolayer studies.
  • Aids solubilization of membrane proteins and hydrophobic compounds.
  • Supports chiral separations of certain herbicides and amino acid derivatives.
  • High listed purity (≈99.8%) for reproducible results.
  • Molecular weight 306.40 and formula C15H30O6.
  • Available in solid and solution formats for flexible use.