Learn More
Medchemexpress LLC Nonyl β-D-glucopyranoside | 69984-73-2 | MFCD00063300 | 99.8% | 306.40 | C15H30O6 | 500 MG
SDP
Catalog No. 5000302117
Encompass_Preferred
Nonyl β-D-glucopyranoside is a nonionic alkyl glycoside surfactant used as a mild detergent and for studies of lipid monolayers and membrane interactions. It is supplied as a solid and as a 10 mM solution in DMSO, and is suitable for biochemical and membrane protein applications.
  • Nonionic surfactant for lipid monolayer and membrane interaction studies.
  • Mild detergent suitable for membrane protein isolation and biochemical assays.
  • High purity (99.79%) for reproducible experimental results.
  • Available as solid in multiple sizes and as a 10 mM solution in DMSO.
  • Solid appearance simplifies storage and handling for lab use.
Catalog No. 50-003-02117 Supplier Medchemexpress LLC Supplier No. HYW015993500MG
Nonyl β-D-glucopyranoside is a nonionic alkyl glycoside surfactant used as a mild detergent and for studies of lipid monolayers and membrane interactions. It is supplied as a solid and as a 10 mM solution in DMSO, and is suitable for biochemical and membrane protein applications.
  • Nonionic surfactant for lipid monolayer and membrane interaction studies.
  • Mild detergent suitable for membrane protein isolation and biochemical assays.
  • High purity (99.79%) for reproducible experimental results.
  • Available as solid in multiple sizes and as a 10 mM solution in DMSO.
  • Solid appearance simplifies storage and handling for lab use.