Learn More
Medchemexpress LLC Nonyl β-D-glucopyranoside | 69984-73-2 | MFCD00063300 | 99.8% | 306.40 g·mol-1 | C15H30O6 | 100 MG
SDP
Catalog No. 5000303631
Encompass_Preferred
Nonyl β-D-glucopyranoside is a non-ionic surfactant used to solubilize membrane components and study lipid-detergent interactions. Supplied as a solid with high analytical purity, it forms micelles suitable for membrane biophysics, protein solubilization, and formulation studies.
  • High purity (99.8%).
  • Solid form for straightforward storage and handling.
  • Readily forms micelles for membrane and lipid interaction studies.
  • Effective for solubilization of hydrophobic compounds and membrane proteins.
  • Available in multiple pack sizes for small-scale and bulk experiments.
  • Molecular weight 306.40 g·mol-1.
Catalog No. 50-003-03631 Supplier Medchemexpress LLC Supplier No. HYW015993100MG
Nonyl β-D-glucopyranoside is a non-ionic surfactant used to solubilize membrane components and study lipid-detergent interactions. Supplied as a solid with high analytical purity, it forms micelles suitable for membrane biophysics, protein solubilization, and formulation studies.
  • High purity (99.8%).
  • Solid form for straightforward storage and handling.
  • Readily forms micelles for membrane and lipid interaction studies.
  • Effective for solubilization of hydrophobic compounds and membrane proteins.
  • Available in multiple pack sizes for small-scale and bulk experiments.
  • Molecular weight 306.40 g·mol-1.