Learn More
Medchemexpress LLC Nonyl β-D-glucopyranoside | 69984-73-2 | MFCD00063300 | 99.8% | 306.40 g·mol⁻1 | C15H30O6 | 250 MG
SDP
Catalog No. 5000302446
Encompass_Preferred
Nonyl β-D-glucopyranoside is a non-ionic alkyl glucoside surfactant used in biochemical and biophysical research. Supplied as a solid (white to off-white) with high purity, it is appropriate for studies of lipid monolayer interactions, membrane-related work, and for use in enantiomer separations of certain herbicides and amino acid derivatives.
  • High purity (99.79%) suitable for analytical and preparative work.
  • Non-ionic surfactant effective for membrane and surface studies.
  • Solid, white to off-white appearance for easy handling and storage.
  • Useful for studies of lipid monolayer interactions.
  • Facilitates enantiomer separation of phenoxy acid herbicides and amino acid derivatives.
  • Molecular formula C15H30O6; molecular weight 306.40.
Catalog No. 50-003-02446 Supplier Medchemexpress LLC Supplier No. 250MGHYW015993
Nonyl β-D-glucopyranoside is a non-ionic alkyl glucoside surfactant used in biochemical and biophysical research. Supplied as a solid (white to off-white) with high purity, it is appropriate for studies of lipid monolayer interactions, membrane-related work, and for use in enantiomer separations of certain herbicides and amino acid derivatives.
  • High purity (99.79%) suitable for analytical and preparative work.
  • Non-ionic surfactant effective for membrane and surface studies.
  • Solid, white to off-white appearance for easy handling and storage.
  • Useful for studies of lipid monolayer interactions.
  • Facilitates enantiomer separation of phenoxy acid herbicides and amino acid derivatives.
  • Molecular formula C15H30O6; molecular weight 306.40.