Learn More
Medchemexpress LLC Nonyl β-D-glucopyranoside | 69984-73-2 | MFCD00063300 | 99.8% | 306.40 g·mol⁻¹ | C15H30O6 | 1 ML
SDP
Catalog No. 5000299933
Encompass_Preferred
Nonyl β-D-glucopyranoside is a non-ionic alkylglucoside surfactant provided as a 10 mM solution in DMSO for laboratory research. It is commonly used to solubilize membrane proteins, investigate lipid monolayer interactions, and support enantiomer separation in biophysical and biochemical assays, with specifications that support reproducible experimental use.
  • High purity for reproducible results.
  • Ready-to-use 10 mM solution in DMSO.
  • Non-ionic surfactant suitable for membrane protein solubilization.
  • Useful for lipid monolayer and biophysical studies.
  • Stable under recommended storage conditions for extended usability.
Catalog No. 50-002-99933 Supplier Medchemexpress LLC Supplier No. HYW01599310MM/1ML
Nonyl β-D-glucopyranoside is a non-ionic alkylglucoside surfactant provided as a 10 mM solution in DMSO for laboratory research. It is commonly used to solubilize membrane proteins, investigate lipid monolayer interactions, and support enantiomer separation in biophysical and biochemical assays, with specifications that support reproducible experimental use.
  • High purity for reproducible results.
  • Ready-to-use 10 mM solution in DMSO.
  • Non-ionic surfactant suitable for membrane protein solubilization.
  • Useful for lipid monolayer and biophysical studies.
  • Stable under recommended storage conditions for extended usability.