Learn More
Cayman Chemical Nonyloxytryptamine (oxalate), Purity: ≥98%, CAS: 157798-13-5, 10 mg
SDP
Catalog No. 502986439
Encompass_Preferred
A potent 5-HT1Dβ receptor agonist (EC50 = 68 nM for inhibition of adenylate cyclase activity); selective for 5-HT1Dβ over 5-HT1A for its adenylate cyclase activity (EC50 = >1 μM for 5-HT1A; binds to 5-HT1Dβ, 5-HT1A, 5-HT1B, 5-HT2A, and 5-HT2C receptors in vitro (Kis = 1.2-315 nM); inhibits reovirus-mediated cell death in a dose-dependent manner but has no effect on reovirus attachment or internalization
Catalog No. 50-298-6439 Supplier Cayman Chemical Supplier No. 2151710 MG
A potent 5-HT1Dβ receptor agonist (EC50 = 68 nM for inhibition of adenylate cyclase activity); selective for 5-HT1Dβ over 5-HT1A for its adenylate cyclase activity (EC50 = >1 μM for 5-HT1A; binds to 5-HT1Dβ, 5-HT1A, 5-HT1B, 5-HT2A, and 5-HT2C receptors in vitro (Kis = 1.2-315 nM); inhibits reovirus-mediated cell death in a dose-dependent manner but has no effect on reovirus attachment or internalization