Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nonylphenol

Catalog No. 29085825ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 L
3 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-085-825ML 25 mL
29-085-81L 1 L
29-085-83L 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 29-085-825ML Supplier Sigma Aldrich Supplier No. 29085825ML
Add to Cart
Add to Cart
This item is not returnable. View return policy

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Nonylphenol is a toxic xenobiotic compound classified as an endocrine disrupter capable of interfering with the hormonal system of numerous organisms
  • Nonylphenol has been reported as antiandrogen by using a yeast detection system
  • Molecular mechanism for the antiandrogenic action of nonylphenol has been reported

Specifications

CAS 84852-15-3
Density 0.937 g/mL (at 25°C (literature))
Molecular Formula C15H24O
Refractive Index n20/D 1.511 (literature)
Linear Formula C15H24O
Quantity 25 mL
Molecular Weight (g/mol) 220.35
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.