Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonylphenol diethoxylate Solution, 50 μg/mL in Acetone, MilliporeSigma™ Supelco™

Catalog No. 111016621 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6621 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6621 Supplier MilliporeSigma Supelco Supplier No. 328981ML
Add to Cart
Add to Cart

Nonylphenol diethoxylate (50 μg/mL in acetone, technical) is classified under the family of alkylphenol ethoxylate surfactants majorly used for households, industry and agriculture.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-64-1
Flash Point -17°C
Linear Formula C19H32O3
Synonym NP-2 EO technical
Type Analytical Standard
Quantity 1 mL
Formula Weight 308.46
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 50 μg/mL in acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.