Learn More
Glass Expansion Noordermeer V-Groove Nebulizer 1L/min Ar flow with Quick Connect Fittings
SDP
Catalog No. 5000556286
Encompass_Preferred
Noordermeer V-Groove Nebulizer 1L/min Ar flow with Quick Connect Fittings Sourced from Glass Expansion
Catalog No. 50-005-56286 Supplier Glass Expansion Supplier No. 018011228
Noordermeer V-Groove Nebulizer 1L/min Ar flow with Quick Connect Fittings Sourced from Glass Expansion