Learn More
Medchemexpress LLC HY-N2195 100mg Medchemexpress, Nootkatone CAS:4674-50-4 Purity:>98%
SDP
Catalog No. 501970375
Encompass_Preferred
Medchemexpress, HY-N2195 100mg Nootkatone CAS:4674-50-4 Nootkatone, a neuroprotective agent from Alpiniae Oxyphyllae Fructus, has antioxidant and anti-inflammatory effects. Nootkatone improves cognitive impairment in lipopolysaccharide-induced mouse model of Alzheimer's disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0375 Supplier Medchemexpress LLC Supplier No. HYN2195100MG
Medchemexpress, HY-N2195 100mg Nootkatone CAS:4674-50-4 Nootkatone, a neuroprotective agent from Alpiniae Oxyphyllae Fructus, has antioxidant and anti-inflammatory effects. Nootkatone improves cognitive impairment in lipopolysaccharide-induced mouse model of Alzheimer's disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.