Learn More
Apexbio Technology LLC Nootkatone 4674-50-4 200mg
SDP
Catalog No. 5000096415
Encompass_Preferred
Nootkatone (CAS nan) is a naturally derived agent investigated for its neuroprotective effects It is designed to modulate intracellular signaling pathways and enzymatic activities associated with neuroinflammation and oxidative stress thereby regulating neuronal cellular processes Nootkatone exerts its biological activity primarily through modulation of signaling pathways and enzymatic activities linked to neuroinflammation and oxidative stress In animal models specifically lipopolysaccharide (LPS)-induced Alzheimer s disease in mice Nootkatone demonstrates amelioration of cognitive impairment Based on these pharmacological properties Nootkatone holds research potential in neuronal protection cognitive function restoration and elucidation of mechanisms related to neurodegenerative pathology
Catalog No. 50-000-96415 Supplier Apexbio Technology LLC Supplier No. C6986200
Nootkatone (CAS nan) is a naturally derived agent investigated for its neuroprotective effects It is designed to modulate intracellular signaling pathways and enzymatic activities associated with neuroinflammation and oxidative stress thereby regulating neuronal cellular processes Nootkatone exerts its biological activity primarily through modulation of signaling pathways and enzymatic activities linked to neuroinflammation and oxidative stress In animal models specifically lipopolysaccharide (LPS)-induced Alzheimer s disease in mice Nootkatone demonstrates amelioration of cognitive impairment Based on these pharmacological properties Nootkatone holds research potential in neuronal protection cognitive function restoration and elucidation of mechanisms related to neurodegenerative pathology