Learn More
Medchemexpress LLC Nootkatone | 4674-50-4 | MFCD00036591 | 250 MG
SDP
Catalog No. 5000421925
Encompass_Preferred
Nootkatone, a neuroprotective agent derived from *Vitis vinifera*, exhibits antioxidant and anti-inflammatory effects, improving cognitive impairment in models of Alzheimer's disease. It also acts as an effective repellent and insecticide against various insects and shows efficacy against *Staphylococcus aureus* biofilms, making it useful in a range of applications.
  • Exhibits neuroprotective, antioxidant, and anti-inflammatory properties.
  • Improves cognitive impairment in Alzheimer's disease models.
  • Effective repellent and insecticide against mosquitos, ticks, and other insects.
  • Demonstrates efficacy against *Staphylococcus aureus* biofilms.
  • Inhibits platelet aggregation and reduces plasma glucose levels.
  • For research use only.
Catalog No. 50-004-21925 Supplier Medchemexpress LLC Supplier No. 250MGHYN2195
Nootkatone, a neuroprotective agent derived from *Vitis vinifera*, exhibits antioxidant and anti-inflammatory effects, improving cognitive impairment in models of Alzheimer's disease. It also acts as an effective repellent and insecticide against various insects and shows efficacy against *Staphylococcus aureus* biofilms, making it useful in a range of applications.
  • Exhibits neuroprotective, antioxidant, and anti-inflammatory properties.
  • Improves cognitive impairment in Alzheimer's disease models.
  • Effective repellent and insecticide against mosquitos, ticks, and other insects.
  • Demonstrates efficacy against *Staphylococcus aureus* biofilms.
  • Inhibits platelet aggregation and reduces plasma glucose levels.
  • For research use only.