Learn More
ABclonal Technology NOP16 Rabbit pAb
SDP
Supplier:  ABclonal Technology A136971000UL
Encompass_Preferred
This gene encodes a protein that is localized to the nucleolus. Expression of this gene is induced by estrogens and Myc protein and is a marker of poor patient survival in breast cancer. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-15203