Learn More
Eurofins Discoverx ChemiScreen NOP/ORL1 Opioid Receptor Membrane Preparation, 1 vial. (HTS040M)
Catalog No. 502005730
Encompass
The ChemiScreen NOP/ORL1 Opioid Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of OPRL1 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.
Catalog No. 50-200-5730 Supplier Eurofins Discoverx Supplier No. HTS040M
The ChemiScreen NOP/ORL1 Opioid Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of OPRL1 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.