Learn More
ABclonal Technology NOP58 Rabbit pAb
SDP
Catalog No. 5000108433
Encompass_Preferred
The protein encoded by this gene is a core component of box C/D small nucleolar ribonucleoproteins. Some box C/D small nucleolar RNAs (snoRNAs), such as U3, U8, and U14, are dependent upon the encoded protein for their synthesis. This protein is SUMOylated, which is necessary for high affinity binding to snoRNAs.
Catalog No. 50-001-08433 Supplier ABclonal Technology Supplier No. A141291000UL
Add to Cart
Add to Cart
The protein encoded by this gene is a core component of box C/D small nucleolar ribonucleoproteins. Some box C/D small nucleolar RNAs (snoRNAs), such as U3, U8, and U14, are dependent upon the encoded protein for their synthesis. This protein is SUMOylated, which is necessary for high affinity binding to snoRNAs.