Learn More
Enzo Life Sciences NOR-3 (5mg). CAS: 138472-01-2
SDP
Supplier:  Enzo Life Sciences ALX430011M005
Encompass_Preferred
Non-thiol-based nitric oxide (NO) donor. NOR-3 shows a variety of biological and pharmacological effects. Strong vasodilatory effects in rat aorta, rabbit aorta and dog coronary artery; potent inhibitory activities against antiplatelet aggregation and thrombus formation; effective aggregation inhibition of human platelet raising by ADP; cardioprotective effects in the ischemia/reperfusion system; antianginal effect. Alternative name: FK409, (+/-)-(E)-4-Ethyl-2-[(E)-hydroxyimino]-5-nitro-3-hexeneamide. Purity: ≥97% (HPLC). Appearance: White to off-white crystalline solid. Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9133