Learn More
Enzo Life Sciences NOR-5 (10mg)
SDP
Supplier:  Enzo Life Sciences ALX430125M010
Encompass_Preferred
Nitric oxide (NO) donor. The half-life time of 0.5mM NOR-5 is 20 hours at 37°C in 0.1M PBS, pH 7.4. Alternative name: (+/-)-2-((E)-4-Ethyl-3[(Z)-hydroxyimino]6-methyl-5-nitro-heptenyl)-3-pyridinecarboxamide. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9213