Learn More
Apexbio Technology LLC nor-Binaltorphimine dihydrochloride 113158-34-2 50mg
SDP
Catalog No. 5000087902
Encompass_Preferred
nor-Binaltorphimine dihydrochloride (CAS 113158-34-2) is a highly selective antagonist of the -opioid receptor By inhibiting -opioid receptor-mediated signaling pathways this compound modulates processes such as nociceptive transmission and regulation of affective states Its selectivity enables precise investigation of -opioid receptor function in experimental models nor-Binaltorphimine dihydrochloride is widely utilized in pain research and neuroscience to elucidate the physiological and behavioral roles of -opioid receptor signaling
Catalog No. 50-000-87902 Supplier Apexbio Technology LLC Supplier No. B626950
nor-Binaltorphimine dihydrochloride (CAS 113158-34-2) is a highly selective antagonist of the -opioid receptor By inhibiting -opioid receptor-mediated signaling pathways this compound modulates processes such as nociceptive transmission and regulation of affective states Its selectivity enables precise investigation of -opioid receptor function in experimental models nor-Binaltorphimine dihydrochloride is widely utilized in pain research and neuroscience to elucidate the physiological and behavioral roles of -opioid receptor signaling