Learn More
Apexbio Technology LLC nor-NOHA (acetate) 1140844-63-8 10mg
SDP
Catalog No. 5000087828
Encompass_Preferred
nor-NOHA (acetate) (CAS 1140844-63-8) is a potent reversible inhibitor of rat liver arginase (Ki 0 5 M) By competitively inhibiting arginase nor-NOHA modulates the cellular balance between the L-arginine-dependent pathways of nitric oxide synthase (NOS) and arginase affecting nitric oxide production and urea cycle metabolism In vitro studies have shown that nor-NOHA suppresses proliferation migration and invasion of HepG2 hepatocellular carcinoma cells induces apoptosis and alters expression of Arg1 MMP-2 P53 and ECD In vivo nor-NOHA improves endothelial function in rat models of inflammation by enhancing NOS activity increasing EDHF and reducing superoxide anion production This compound is widely used to investigate arginase-related mechanisms in cellular and vascular biology
Catalog No. 50-000-87828 Supplier Apexbio Technology LLC Supplier No. C540710
nor-NOHA (acetate) (CAS 1140844-63-8) is a potent reversible inhibitor of rat liver arginase (Ki 0 5 M) By competitively inhibiting arginase nor-NOHA modulates the cellular balance between the L-arginine-dependent pathways of nitric oxide synthase (NOS) and arginase affecting nitric oxide production and urea cycle metabolism In vitro studies have shown that nor-NOHA suppresses proliferation migration and invasion of HepG2 hepatocellular carcinoma cells induces apoptosis and alters expression of Arg1 MMP-2 P53 and ECD In vivo nor-NOHA improves endothelial function in rat models of inflammation by enhancing NOS activity increasing EDHF and reducing superoxide anion production This compound is widely used to investigate arginase-related mechanisms in cellular and vascular biology