Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nor-NOHA acetate | 1140844-63-8 | ≥99.0% | 296.28 g/mol | C9H20N4O7 | 5 MG
SDP

Catalog No. 5000262438
Encompass_Preferred

nor-NOHA acetate (Nω-hydroxy-nor-L-arginine acetate) is a research-grade, selective and reversible arginase inhibitor used to study endothelial dysfunction, immunosuppression, apoptosis, and metabolic pathways. Supplied as the acetate salt in solid and solution formats for laboratory research; intended for in vitro studies.

  • Selective and reversible arginase inhibitor.
  • Induces apoptosis in ARG2-expressing cells under hypoxia; useful in endothelial dysfunction and immunosuppression research.
  • Supplied as acetate salt in solid (1 mg, 5 mg, 10 mg, 50 mg) and solution (10 mM in water, 1 mL) formats.
  • High purity (≥99.0%).
  • Molecular weight 296.28 g/mol; molecular formula C9H20N4O7.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-62438 Supplier Medchemexpress LLC Supplier No. HY112885A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

nor-NOHA acetate (Nω-hydroxy-nor-L-arginine acetate) is a research-grade, selective and reversible arginase inhibitor used to study endothelial dysfunction, immunosuppression, apoptosis, and metabolic pathways. Supplied as the acetate salt in solid and solution formats for laboratory research; intended for in vitro studies.

  • Selective and reversible arginase inhibitor.
  • Induces apoptosis in ARG2-expressing cells under hypoxia; useful in endothelial dysfunction and immunosuppression research.
  • Supplied as acetate salt in solid (1 mg, 5 mg, 10 mg, 50 mg) and solution (10 mM in water, 1 mL) formats.
  • High purity (≥99.0%).
  • Molecular weight 296.28 g/mol; molecular formula C9H20N4O7.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.