Learn More
Enzo Life Sciences nor-NOHA . acetate, (1mg), CAS Number: 189302-40-7
SDP
Catalog No. NC1369639
Encompass_Preferred
Arginase inhibitor. Potent and reversible arginase inhibitor. Specifically interacts with the manganese-cluster of the arginase active site. Does not act as a substrate or as an inhibitor of NOS. Alternative Name: Nomega-Hydroxy-nor-L-arginine. diacetate. Formula: C5H12N4O3. CH3COOH. MW: 176.2. 60.1. CAS Number: 189302-40-7. Purity: ≥97% (TLC). Appearance: White to slightly yellowish powder. Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C
Catalog No. NC1369639 Supplier Enzo Life Sciences Supplier No. ALX270467M001
Arginase inhibitor. Potent and reversible arginase inhibitor. Specifically interacts with the manganese-cluster of the arginase active site. Does not act as a substrate or as an inhibitor of NOS. Alternative Name: Nomega-Hydroxy-nor-L-arginine. diacetate. Formula: C5H12N4O3. CH3COOH. MW: 176.2. 60.1. CAS Number: 189302-40-7. Purity: ≥97% (TLC). Appearance: White to slightly yellowish powder. Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C