Learn More
Enzo Life Sciences nor-NOHA. acetate (25mg). CAS: 189302-40-7
SDP
Catalog No. 502008592
Encompass_Preferred
Potent and reversible arginase inhibitor. Specifically interacts with the manganese-cluster of the arginase active site. Does not act as a substrate or as an inhibitor of NOS. Alternative name: Nomega-Hydroxy-nor-L-arginine . diacetate. Purity: ≥97% (TLC). Appearance: White to slightly yellowish powder. Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8592 Supplier Enzo Life Sciences Supplier No. ALX270467M025
Potent and reversible arginase inhibitor. Specifically interacts with the manganese-cluster of the arginase active site. Does not act as a substrate or as an inhibitor of NOS. Alternative name: Nomega-Hydroxy-nor-L-arginine . diacetate. Purity: ≥97% (TLC). Appearance: White to slightly yellowish powder. Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.