Learn More
Medchemexpress LLC nor-NOHA monoacetate | 2250019-93-1 | 99.9% | 236.23 | 100 MG
SDP
Catalog No. 5000438778
Encompass_Preferred
nor-NOHA monoacetate is a selective and reversible arginase inhibitor. It induces apoptosis in ARG2-expressing cells under hypoxia and exhibits anti-leukemic activity. It can be utilized in studies concerning endothelial dysfunction, immunosuppression, and metabolism.
  • Selective and reversible arginase inhibitor
  • Induces apoptosis in ARG2-expressing cells under hypoxia
  • Exhibits anti-leukemic activity
  • Can be used in studies of endothelial dysfunction, immunosuppression, and metabolism
  • Induces apoptosis in a dose-dependent manner in K562 cells under hypoxia
  • Attenuates hypoxia-mediated resistance towards imatinib in K562 or KCL22 cells
  • Induces a significant reduction of infarct size in male Sprague-Dawley rats
  • Increases plasma citrulline and nitrite levels and decreases ornithine plasma levels in male Sprague-Dawley rats
Catalog No. 50-004-38778 Supplier Medchemexpress LLC Supplier No. HY112885B100MG
nor-NOHA monoacetate is a selective and reversible arginase inhibitor. It induces apoptosis in ARG2-expressing cells under hypoxia and exhibits anti-leukemic activity. It can be utilized in studies concerning endothelial dysfunction, immunosuppression, and metabolism.
  • Selective and reversible arginase inhibitor
  • Induces apoptosis in ARG2-expressing cells under hypoxia
  • Exhibits anti-leukemic activity
  • Can be used in studies of endothelial dysfunction, immunosuppression, and metabolism
  • Induces apoptosis in a dose-dependent manner in K562 cells under hypoxia
  • Attenuates hypoxia-mediated resistance towards imatinib in K562 or KCL22 cells
  • Induces a significant reduction of infarct size in male Sprague-Dawley rats
  • Increases plasma citrulline and nitrite levels and decreases ornithine plasma levels in male Sprague-Dawley rats