Learn More
Medchemexpress LLC HY-112885B 5mg Medchemexpress, nor-NOHA (monoacetate) CAS:2250019-93-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112885B5MG
Encompass_Preferred
Medchemexpress, HY-112885B 5mg nor-NOHA (monoacetate) CAS:2250019-93-1 nor-NOHA (Nω-Hydroxy-nor-L-arginine) monoacetate is a potent and selective arginase inhibitor. nor-NOHA monoacetate inhibits rat liver arginase with a Ki of 0.5 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0827