Learn More
Cayman Chemical nor-W-18-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2701349-29-1, 1 mg
SDP
Catalog No. 502983999
Encompass_Preferred
An internal standard for the quantification of nor-W-18 by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-298-3999 Supplier Cayman Chemical Supplier No. 203171 MG
An internal standard for the quantification of nor-W-18 by GC- or LC-MS; intended for research and forensic applications